Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Rituximab, Clone: 109H9.06, Novus Biologicals™

Mouse Monoclonal Antibody

Specifications

Antigen Rituximab
Clone 109H9.06
Concentration 0.5mg/mL
Immunogen Anti-Mabthera(Rituximab)
Formulation PBS and 50% Glycerol with No Preservative
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
DDX5010B05
View Documents
Novus Biologicals
DDX5010B1000.05MG
0.05 mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
DDX5010P05
View Documents
Novus Biologicals
DDX5010P1000.05MG
0.05 mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Rituximab Monoclonal antibody specifically detects Rituximab in Human samples. It is validated for ELISA.
Specifications

Specifications

Rituximab
0.5mg/mL
PBS and 50% Glycerol with No Preservative
Primary
Mouse
Biotin
Ion exchange chromatography
Western Blot
109H9.06
Anti-Mabthera(Rituximab)
Monoclonal
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Purified
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit