Learn More
Mouse anti-RNASEH2B, Clone: 419, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243774
Catalog No. NBP243774
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot
RNASEH2B Monoclonal antibody specifically detects RNASEH2B in Human samples. It is validated for Western Blot.
Specifications
Specifications
RNASEH2B
419
Western Blot 1:500-1:3000
Purified
RNASEH2B
Recombinant protein encompassing a sequence within the center region of human RNaseH2B. The exact sequence is proprietary.
35 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot
Unconjugated
PBS with No Preservative
ribonuclease H2, subunit B
Mouse
IgG1
Protein G purified
RUO
Primary
79621