Learn More
RNF11, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant RNF11.
Supplier: Abnova Corporation H00026994A01
Description
The protein encoded by this gene contains a RING-H2 finger motif, which is known to be important for protein-protein interactions. The expression of this gene has been shown to be induced by mutant RET proteins (MEN2A/MEN2B). The germline mutations in RET gene are known to be responsible for the development of multiple endocrine neoplasia (MEN). [provided by RefSeq
Sequence: EEQIRIAQRIGLIQHLPKGVYDPGRDGSEKKIRECVICMMDFVYGDPIRFLPCMHIYHLDCIDDWLMRSFTCPSCMEPVDAALLSSYETNSpecifications
RNF11 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant RNF11. | |
RNF11 | |
CGI-123/MGC51169/SID1669 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_014372 | |
RNF11 | |
RNF11 (NP_055187, 65 a.a. ∽ 154 a.a) partial recombinant protein with GST tag. | |
RUO | |
26994 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only