Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF180, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant RNF180.
Supplier: Abnova Corporation H00285671A01
Description
Sequence: MKRSKELITKNHSQEETSILRCWKCRKCIASSGCFMEYLENQVIKDKDDSVDAQNICHVWHMNVEALPEWISCLIQKAQWTVGKLNCPFCGARLGGFNFVSpecifications
RNF180 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant RNF180. | |
RNF180 | |
MGC120326/MGC120328 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_178532 | |
RNF180 | |
RNF180 (NP_848627, 1 a.a. ∽ 100 a.a) partial recombinant protein with GST tag. | |
RUO | |
285671 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction