Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-RPL18A, Clone: 6G6G10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237396

Catalog No. NBP237396


Add to cart

Description

Description

RPL18A Monoclonal antibody specifically detects RPL18A in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications

Specifications

RPL18A
6G6G10
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
Ascites
RPL18A
Purified recombinant fragment of human RPL18A (AA: 50-176) expressed in E. Coli.
20.8 kDa
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Ascites with 0.03% Sodium Azide
60S ribosomal protein L18a, ribosomal protein L18a, ribosomal protein L18a-like protein
Mouse
IgG1
Unpurified
RUO
Primary
6142
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only