Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RRBP1 Antibody (5610), Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP243600
Description
Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence
RRBP1 Monoclonal specifically detects RRBP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
RRBP1 | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
RRBP1 | |
Recombinant protein encompassing a sequence within the C-terminus region of human Ribosome binding protein 1. The exact sequence is proprietary. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence, Proximity Ligation Assay | |
5610 | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:2000, Proximity Ligation Assay | |
180 kDa ribosome receptor homolog, DKFZp586A1420, ES/130, ES/130-related protein, ES130, FLJ36146, hES, KIAA1398, MGC157720, MGC157721, ribosome binding protein 1 (dog 180kD homolog), ribosome binding protein 1 homolog 180kDa (dog), Ribosome receptor protein, ribosome-binding protein 1, RRp | |
Mouse | |
Protein G purified | |
RUO | |
6238 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction