Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-RSK2, Clone: 4E10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237272

Catalog No. NBP237272


Add to cart

Description

Description

RSK2 Monoclonal antibody specifically detects RSK2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

RSK2
40000000000
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000
Ascites
RPS6KA3
Purified recombinant fragment of human RSK2 expressed in E. Coli.
80 kDa
0.1 ml
Cancer, Protein Kinase
Primary
6197
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
Ascites with 0.03% Sodium Azide
90 kDa ribosomal protein S6 kinase 3, CLS, Coffin-Lowry syndrome, EC 2.7.11, EC 2.7.11.1, HU-3, Insulin-stimulated protein kinase 1, ISPK1, ISPK-1, MAP kinase-activated protein kinase 1b, MAPK-activated protein kinase 1b, MAPKAP kinase 1b, MAPKAPK-1b, MAPKAPK1B, mental retardation, X-linked 19, MRX19, p90-RSK2, p90RSK3, pp90RSK2, ribosomal protein S6 kinase alpha-3, ribosomal protein S6 kinase, 90kD, polypeptide 3, ribosomal protein S6 kinase, 90kDa, polypeptide 3, Ribosomal S6 kinase 2, RSK, RSK-2, RSK2p90-RSK 3, S6K-alpha3, S6K-alpha-3
Mouse
IgG1
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only