Learn More
SAA, Mouse, Clone: 291, Abnova™
Mouse monoclonal antibody raised against recombinant SAA.
Manufacturer:  Abnova Corporation MAB6410
Catalog No. 89-136-200
Add to cart
Description
Description
Specifications
Specifications
SAA
291
Mouse monoclonal antibody raised against recombinant SAA.
Lyophilized from 10mM PBS, pH 7.2
Mouse
IgG2b, kappa
RUO
Primary
This antibody reacts with natural and recombinant human SAA.
ELISA, Western Blot
Unconjugated
Western Blot (0.1-0.5 ug) The optimal working dilution should be determined by the end user.
Lyophilized
Recombinant protein corresponding to human Serum Amyloid A (SAA).
500 ug
Store at -20°C on dry atmosphere. After reconstitution with deionized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human