Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAP102 Mouse anti-Mouse, Rat, Clone: 7D3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP1974670.2MG
Description
SAP102 Monoclonal specifically detects SAP102 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
SAP102 | |
Monoclonal | |
Unconjugated | |
PBS and 50% Glycerol with 0.09% Sodium Azide | |
DLG3 | |
Recombinant rat SAP102 (N-terminal fragment). | |
0.2 mg | |
Cell Cycle and Replication, Neuroscience | |
1741 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
7D3 | |
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
discs, large homolog 3 (Drosophila), KIAA1232discs, large homolog 3 (neuroendocrine-dlg, Drosophila), MRX90MRX, NEDLGdisks large homolog 3, neuroendocrine-DLG, SAP-102, SAP102NE-Dlg, Synapse-associated protein 102, XLMR | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction