Learn More
Mouse anti-Seasonal H1N1 Neuraminidase, Clone: 10C5F12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP241331
Catalog No. NBP241331
Add to cart
Description
Description
Seasonal H1N1 Neuraminidase Monoclonal antibody specifically detects Seasonal H1N1 Neuraminidase in Virus samples. It is validated for ELISA.
Specifications
Specifications
Seasonal H1N1 Neuraminidase
10C5F12
Unconjugated
PBS with 0.02% Sodium Azide
Mouse
IgG2b
0.1 mg
Store at -20 degrees C. Avoid freeze/thaw cycles.
Monoclonal
ELISA
1 mg/mL
ELISA
Affinity Purified
Antibody was raised against a synthetic peptide from the seasonal influenza (H1N1) neuraminidase protein.
Protein A purified
RUO
Primary
Virus