Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Semaphorin 4D/CD100, Clone: SEMA4D/933, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP2443640.1MG

 View more versions of this product

Catalog No. NBP24436401


Add to cart

Description

Description

Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence

Semaphorin 4D/CD100 Monoclonal antibody specifically detects Semaphorin 4D/CD100 in Human, Primate samples. It is validated for Flow Cytometry, Immunofluorescence.
Specifications

Specifications

Semaphorin 4D/CD100
SEMA4D/933
Unconjugated
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Purified
SEMA4D
Human recombinant SEMA4D protein
Protein A or G purified
RUO
Store at 4C.
Monoclonal
Recognizes a homodimeric protein comprised of 50kDa subunits, identified as CD100. It is expressed on majority of haemopoietic cells (B, T, NK and myeloid cells) and is absent from bone marrow, erythrocytes, eosinophils and endothelial cells. Its expression is increased after PHA-activation. CD100 was shown to associate with different partner molecules in T cells such as CD45, a key molecule with protein tyrosine phosphatase activity involved in T-cell transduction, and a Serine kinase. It plays a role in homotypic cell adhesion and in T cell activation.
Flow Cytometry, Immunofluorescence
0.2 mg/mL
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml
Q92854
A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D
Mouse
IgG1 Kappa
0.1 mg
Apoptosis, Immunology
Primary
10507
Human, Primate
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only