Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Serpin A7/TBG, Clone: 1C3H11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237323

Catalog No. NBP237323


Add to cart

Description

Description

Serpin A7/TBG Monoclonal antibody specifically detects Serpin A7/TBG in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Serpin A7/TBG
1C3H11
Unconjugated
PBS with 0.05% Sodium Azide
antitrypsin), member 7, FLJ17820, member 7, Serpin A7, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), T4-binding globulin, TBGclade A (alpha-1 antiproteinase, thyroxin-binding globulin, thyroxine-binding globulin
Mouse
IgG1
Protein A or G purified
RUO
Primary
6906
Western Blot, ELISA
1 mg/mL
Western Blot 1:500 - 1:2000, ELISA 1:10000
Affinity Purified
SERPINA7
Purified recombinant fragment of human SERPINA7 (AA: 168-302) expressed in E. Coli.
46.3 kDa
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only