Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Siglec-2/CD22, Clone: FR10B4, Novus Biologicals™

Mouse Monoclonal Antibody

$129.00 - $299.00

Specifications

Antigen Siglec-2/CD22
Clone FR10B4
Concentration 0.2 mg/mL
Immunogen Human Burkitt's lymphoma cells
Gene ID (Entrez) 933
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP24460100 View Documents Novus BiologicalsSupplier Diversity Partner
NBP2446010.02MG
0.02 mg Each for $129.00
Add to cart
 
NBP24460101 View Documents Novus BiologicalsSupplier Diversity Partner
NBP2446010.1MG
0.1 mg Each for $299.00
Add to cart
 
NBP24460102 View Documents Novus BiologicalsSupplier Diversity Partner
NBP2446010.2MG
0.2 mg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence

Siglec-2/CD22 Monoclonal antibody specifically detects Siglec-2/CD22 in Human samples. It is validated for Flow Cytometry, Immunofluorescence.
Specifications

Specifications

Siglec-2/CD22
0.2 mg/mL
933
Monoclonal
Mouse
IgG1 Kappa
Purified
RUO
Protein A or G purified
FR10B4
Human Burkitt's lymphoma cells
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Primary
Recognizes a protein of 130-140kDa, identified as CD22 (also known as BL-CAM). CD22 expression is restricted to normal and neoplastic B cells and is absent from other haemopoietic cell types. In B-cell ontogeny, CD22 is first expressed in the cytoplasm of pro-B and pre-B cells, and on the surface as B cells mature to become IgD+. It is not expressed by plasma cells, CD22 is found highly expressed in follicular mantle and marginal zone B-cells, and while germinal center B-cells are relatively weak. CD22 is a member of the immunoglobulin superfamily and serves as an adhesion receptor for sialic acid-bearing ligands expressed on erythrocytes and all leukocyte classes. It also associates with tyrosine kinases and play a role in signal transduction and B-cell activation.
Unconjugated
Flow Cytometry, Immunofluorescence
Store at 4C.

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit