Learn More
Mouse anti-SLC31A1/CTR1, Alexa Fluor 647, Clone: 5E6.1B4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236573AF647
Catalog No. NBP236573Y
Add to cart
Description
Description
SLC31A1/CTR1 Monoclonal antibody specifically detects SLC31A1/CTR1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
SLC31A1/CTR1
5E6.1B4
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
O15431
Copper transporter 1, COPT1high affinity copper uptake protein 1, CTR1MGC75487, hCTR1copper transport 1 homolog, solute carrier family 31 (copper transporters), member 1, Solute carrier family 31 member 1
Mouse
IgG2b Kappa
0.1 ml
Neuroscience, Neurotransmission, Protein Kinase
Primary
1317
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Purified
SLC31A1
Two synthetic peptides made to internal sequences of human SLC31A1/CTR1 protein (sequences found between amino acids 50-150) [UniProt# O15431]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human