Learn More
Mouse anti-SLC31A1/CTR1, HRP, Clone: 5E6.1B4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236573H
Catalog No. NBP236573H
Add to cart
Description
Description
SLC31A1/CTR1 Monoclonal antibody specifically detects SLC31A1/CTR1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
SLC31A1/CTR1
5E6.1B4
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
O15431
Copper transporter 1, COPT1high affinity copper uptake protein 1, CTR1MGC75487, hCTR1copper transport 1 homolog, solute carrier family 31 (copper transporters), member 1, Solute carrier family 31 member 1
Mouse
IgG2b Kappa
0.1 ml
Neuroscience, Neurotransmission, Protein Kinase
Primary
1317
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
HRP
PBS with No Preservative
Purified
SLC31A1
Two synthetic peptides made to internal sequences of human SLC31A1/CTR1 protein (sequences found between amino acids 50-150) [UniProt# O15431]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human