Learn More
SLCO1B1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SLCO1B1.
Supplier: Abnova Corporation H00010599A01
Description
This gene encodes a liver-specific member of the organic anion transporter family. The encoded protein is a transmembrane receptor that mediates the sodium-independent uptake of numerous endogenous compounds including bilirubin, 17-beta-glucuronosyl estradiol and leukotriene C4. This protein is also involved in the removal of drug compounds such as statins, bromosulfophthalein and rifampin from the blood into the hepatocytes. Polymorphisms in the gene encoding this protein are associated with impaired transporter function. [provided by RefSeq
Sequence: YISPCLAGCKSSSGNKKPIVFYNCSCLEVTGLQNRNYSAHLGECPRDDACTRKFYFFVAIQVLNLFFSALGGTSHVMLIVKIVQPELKSLSpecifications
SLCO1B1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant SLCO1B1. | |
SLCO1B1 | |
LST-1/LST1/MGC133282/OATP-C/OATP1B1/OATP2/OATPC/SLC21A6 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_006446 | |
SLCO1B1 | |
SLCO1B1 (NP_006437, 481 a.a. ∽ 570 a.a) partial recombinant protein with GST tag. | |
RUO | |
10599 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only