Learn More
Mouse anti-SOD2/Mn-SOD, Clone: 582, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243795
Catalog No. NBP243795
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot
SOD2/Mn-SOD Monoclonal antibody specifically detects SOD2/Mn-SOD in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
SOD2/Mn-SOD
582
Western Blot 1:1000-1:20000
Purified
SOD2
Recombinant protein encompassing a sequence within the center region of human SOD2. The exact sequence is proprietary.
25 kDa
0.1 ml
Cancer, Diabetes Research, Neuroscience, Tumor Suppressors
Primary
6648
Western Blot
Unconjugated
PBS with No Preservative
EC 1.15.1.1, indophenoloxidase B, IPOB, manganese-containing superoxide dismutase, mangano-superoxide dismutase, Mn superoxide dismutase, MNSOD, MVCD6, superoxide dismutase [Mn], mitochondrial, superoxide dismutase 2, mitochondrial
Mouse
IgG2a
Protein G purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat