Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD3, Mouse, Clone: 1H12, Abnova™

Mouse monoclonal antibody raised against partial recombinant SOD3.

Manufacturer:  Abnova Corporation MAB0749

Catalog No. 89-108-100


Add to cart

Description

Description

This gene encodes a member of the superoxide dismutase (SOD) protein family. SODs are antioxidant enzymes that catalyze the dismutation of two superoxide radicals into hydrogen peroxide and oxygen. The product of this gene is thought to protect the brain, lungs, and other tissues from oxidative stress. The protein is secreted into the extracellular space and forms a glycosylated homotetramer that is anchored to the extracellular matrix (ECM) and cell surfaces through an interaction with heparan sulfate proteoglycan and collagen. A fraction of the protein is cleaved near the C-terminus before secretion to generate circulating tetramers that do not interact with the ECM. [provided by RefSeq

Specifications

Specifications

SOD3
1H12
Mouse monoclonal antibody raised against partial recombinant SOD3.
In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
Liquid
SOD3
Recombinant protein corresponding to amino acids 19-240 of human SOD3.
Ammonium sulfate precipitation
RUO
Primary
6649
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
Western Blot (1:1800) Immunoprecipitation (1 ul) The optimal working dilution should be determined by the end user.
SOD3
EC-SOD/MGC20077
Mouse
IgG2b, kappa
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit