Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Somatostatin Antibody (7G5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP237447
Description
Somatostatin Monoclonal specifically detects Somatostatin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence, CyTOF-ready.Specifications
Somatostatin | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
Growth hormone release-inhibiting factor, SMST, somatostatin, somatostatin-14, somatostatin-28 | |
Mouse | |
12.7 kDa | |
0.1 mL | |
Breast Cancer, Cancer, Stem Cell Markers | |
6750 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
7G5 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
SST | |
Purified recombinant fragment of human SST (AA: 1-116) expressed in E. Coli. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction