Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPDEF, Mouse, Clone: 3, Abnova™
Mouse monoclonal antibody raised against full-length recombinant SPDEF.
Supplier: Abnova Corporation MAB9916
Description
PDEF is an ETS transcription factor expressed in prostate epithelial cells. It acts as an androgen-independent transactivator of PSA (MIM 176820) expression.[supplied by OMIM
Specifications
SPDEF | |
Monoclonal | |
Unconjugated | |
Western blot (1:500) The optimal working dilution should be determined by the end user. | |
SPDEF | |
PDEF/RP11-375E1__A.3/bA375E1.3 | |
Mouse | |
Affinity purification | |
RUO | |
25803 | |
Human | |
Liquid |
ELISA, Western Blot | |
3 | |
Mouse monoclonal antibody raised against full length recombinant SPDEF. | |
In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300) | |
NM_012391.1 | |
SPDEF | |
Recombinant protein corresponding to full length human SPDEF. | |
100 μg | |
Primary | |
It can expression in HeLa whole cell lysate. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction