Learn More
Mouse anti-STAG2, Clone: 1417, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243823
Catalog No. NBP243823
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot
STAG2 Monoclonal antibody specifically detects STAG2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
STAG2
1417
Western Blot 1:500-1:3000
Purified
STAG2
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SA2. The exact sequence is proprietary.
141 kDa
0.1 ml
DNA Repair
Primary
10735
Western Blot
Unconjugated
PBS with No Preservative
bA517O1.1, cohesin subunit SA-2, DKFZp686P168, SA-2, SA2FLJ25871, SCC3 homolog 2, stromal antigen 2DKFZp781H1753
Mouse
IgG2a
Protein A purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Human