Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stomatin Antibody (33F5) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242660

Catalog No. NBP242660


Add to Cart

Description

Description

Ensure accurate, reproducible results in Western Blot, ELISA

Stomatin Monoclonal specifically detects Stomatin in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Stomatin
Monoclonal
1 mg/mL
Western Blot 1:1000, ELISA
NP_004090
STOM
Recombinant human STOM (55-288aa) purified from E. coli
100 μL
Neuroscience
2040
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2b κ
Western Blot, ELISA
33F5
Unconjugated
PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
BND7EPB7, EPB72erythrocyte band 7 integral membrane protein, erythrocyte membrane protein band 7.2 (stomatin), Protein 7.2b, Stomatin
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only