Learn More
Mouse anti-Strep II Tag, Clone: 661, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243719
Catalog No. NBP243719
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Strep II Tag Monoclonal antibody specifically detects Strep II Tag in Non-species specific samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
Specifications
Strep II Tag
661
Unconjugated
PBS with No Preservative
Mouse
IgG2a
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
1mg/mL
Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500
Purified
Immunogen Amino acid sequence - ASWSHPQFEKGA
Protein G purified
RUO
Primary
Non-species specific