Learn More
Mouse anti-T-bet/TBX21, Clone: 4B10, Cy5.5, PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1428350.1MG
 View more versions of this product
Catalog No. NBP14283501
Add to cart
Description
Description
T-bet/TBX21 Monoclonal antibody specifically detects T-bet/TBX21 in Human, Mouse, Rhesus Macaque samples. It is validated for Flow Cytometry.
Specifications
Specifications
T-bet/TBX21
4B10
PerCP-Cyanine5.5
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM
Mouse
IgG1 Kappa
0.1 mg
Store at 4C. Do not freeze.
Monoclonal
Human, Mouse, Rhesus Macaque
Flow Cytometry
0.2 mg/mL
Flow Cytometry 1:10-1:1000
Affinity Purified
TBX21
The immunogen for this antibody was TBet.
Protein A or G purified
RUO
Primary
30009