Learn More
Mouse anti-TCR alpha/beta, Clone: IP26, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1431090.025MG
Catalog No. NBP14310903
Add to cart
Description
Description
TCR alpha/beta Monoclonal antibody specifically detects TCR alpha/beta in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
TCR alpha/beta
IP26
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA
Mouse
IgG1 Kappa
0.025 mg
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary
6955
Flow Cytometry
0.5 mg/mL
Flow Cytometry 1:10-1:1000
Affinity Purified
TRA
The immunogen for this antibody was alpha beta TcR.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human