Learn More
TEAD3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant TEAD3.
Supplier: Abnova Corporation H00007005A01
Description
This gene product is a member of the transcriptional enhancer factor (TEF) family of transcription factors, which contain the TEA/ATTS DNA-binding domain. It is predominantly expressed in the placenta and is involved in the transactivation of the chorionic somatomammotropin-B gene enhancer. Translation of this protein is initiated at a non-AUG (AUA) start codon. [provided by RefSeq
Sequence: WQDRTIASSRLRLLEYSAFMEVQRDPDTYSKHLFVHIGQTNPAFSDPPLEAVDVRQIYDKFPEKKGGLKELYEKGPPNAFFLVKFWADSpecifications
TEAD3 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant TEAD3. | |
TEAD3 | |
DTEF-1/ETFR-1/TEAD5/TEF-5/TEF5 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_003214 | |
TEAD3 | |
TEAD3 (NP_003205, 215 a.a. ∽ 302 a.a) partial recombinant protein with GST tag. | |
RUO | |
7005 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only