Learn More
Mouse anti-TET2, Clone: 649, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243552
Catalog No. NBP243552
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation
TET2 Monoclonal antibody specifically detects TET2 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation.
Specifications
Specifications
TET2
649
Unconjugated
PBS with No Preservative
FLJ20032, MGC125715, tet methylcytosine dioxygenase 2
Mouse
IgG1
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot, ChIP assay, Immunoprecipitation
1mg/mL
Western Blot 1:500-1:3000, Chromatin Immunoprecipitation, Immunoprecipitation 1:100-1:500
Purified
TET2
Recombinant protein encompassing a sequence within the N-terminus region of human TET2. The exact sequence is proprietary.
Protein G purified
RUO
Primary
54790