Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TG, Mouse, Clone: 12G9, Abnova™

Mouse monoclonal antibody raised against native TG.

Manufacturer:  Abnova Corporation MAB0639

Catalog No. 89-108-019

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

TG
12G9
thyroglobulin
TG
AITD3, TGN
Murine
IgG2b
RUO
Primary
7038
Bovine, Canine, Feline, Human, Murine, Porcine, Rat
ELISA
Unconjugated
In PBS, pH 7.4 (0.09% sodium azide)
Liquid
TG
Native purified human TG.
200μL
Store at 4°C.
Monoclonal
This antibody recognizes human TG and do not interfere with anti-thyroglobulin autoantibodies
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit