Learn More
Mouse anti-TGF-alpha, Clone: P/T1, Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP2347020.1 MG
 View more versions of this product
Catalog No. NBP23470201
Description
Description
TGF-alpha Monoclonal antibody specifically detects TGF-alpha in Human, Rabbit, Zebrafish samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications
Specifications
TGF-alpha
P/T1
Unconjugated
10mM PBS - BSA free with No Preservative
Purified
TGFA
A 10-amino acid synthetic peptide (aa 34-43; PPVAAAVVSH) from human TGF alpha.
6 kDa
0.1 mg
Angiogenesis, Cell Cycle and Replication, Signal Transduction
Primary
7039
Protein A purified
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
1.0 mg/mL
Flow Cytometry 0.5-1ug/million cells, ELISA 1-5ug/ml for coating, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug protein, Immunohistochemistry-Paraffin 1-2ug/ml, Immunohistochemistry-Frozen 1-2ug/ml, CyTOF-ready
P01135
protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha
Mouse
IgG1 Kappa
Protein A purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
This antibody reacts with the TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. The staining with this MAb is completely blocked by the peptide used for raising this antibody.
Human, Rabbit, Zebrafish