Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGM2, Mouse, Clone: 2A1, Abnova™

Mouse monoclonal antibody raised against full-length recombinant TGM2.

Manufacturer:  Abnova Corporation MAB0732

Catalog No. 89-108-085


Add to cart

Description

Description

Transglutaminases are enzymes that catalyze the crosslinking of proteins by epsilon-gamma glutamyl lysine isopeptide bonds. While the primary structure of transglutaminases is not conserved, they all have the same amino acid sequence at their active sites and their activity is calcium-dependent. The protein encoded by this gene acts as a monomer, is induced by retinoic acid, and appears to be involved in apoptosis. Finally, the encoded protein is the autoantigen implicated in celiac disease. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Specifications

TGM2
2A1
Mouse monoclonal antibody raised against full length recombinant TGM2.
In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
Liquid
TGM2
Recombinant protein corresponding to full length human TGM2.
Ammonium sulfate precipitation
RUO
Primary
7052
ELISA, Immunoprecipitation, Western Blot
Unconjugated
Western Blot (1:1800) Immunoprecipitation (1-2 ul) The optimal working dilution should be determined by the end user.
TGM2
G-ALPHA-h/GNAH/TG2/TGC
Mouse
IgG1, kappa
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit