Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-TLR2, Clone: TLR2/221, Novus Biologicals™

Mouse Monoclonal Antibody
$244.00 - $488.00
Specifications
Antigen | TLR2 |
---|---|
Clone | TLR2/221 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page, Immunofluorescence 1 - 2 ug/ml |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24516400
![]() |
Novus Biologicals
NBP2451640.02MG |
0.02 mg |
Each for $244.00
|
|
|||||
NBP24516401
![]() |
Novus Biologicals
NBP2451640.1MG |
0.1 mg |
Each for $488.00
|
|
|||||
NBP24516402
![]() |
Novus Biologicals
NBP2451640.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
TLR2 Monoclonal specifically detects TLR2 in Human, Canine, Monkey samples. It is validated for Functional.Specifications
TLR2 | |
0.2 mg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
TLR2 | |
IgG2a κ | |
Protein A or G purified | |
This MAb reacts with human Toll-like receptor 2 (TLR2). It is a member of the Toll-like receptor (TLR) family, which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This gene is expressed most abundantly in peripheral blood leukocytes, and mediates host response to Gram-positive bacteria and yeast via stimulation of NF-kappaB. |
TLR2/221 | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page, Immunofluorescence 1 - 2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Canine, Monkey | |
O60603 | |
7097 | |
Recombinant human TLR2 protein | |
Primary | |
Store at 4C. | |
90 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title