Learn More
Transmissible gastroenteritis virus, Mouse, Clone: TGEV15-948.2.1, Abnova™
Mouse monoclonal antibody raised against Transmissible gastroenteritis virus.
Manufacturer:  Abnova Corporation MAB8327
Catalog No. 89-136-459
Add to cart
Description
Description
Specifications
Specifications
Transmissible gastroenteritis virus
TGEV15-948.2.1
Mouse monoclonal antibody raised against Transmissible gastroenteritis virus.
In PBS, pH 7.4
Mouse
IgG2a
RUO
Primary
These antibodies specifically recognize TGEV and do not cross-react with the following coronaviruses: FIPV, FeCV, CCV.
ELISA, Inhibition Assays
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Native purified from Transmissible gastroenteritis virus-infected ST cells.
100 ug
Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Virus