Learn More
Mouse anti-Triosephosphate isomerase, Clone: 3711, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243825
Catalog No. NBP243825
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot
Triosephosphate isomerase Monoclonal antibody specifically detects Triosephosphate isomerase in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
Triosephosphate isomerase
3711
Western Blot 1:500-1:3000
Purified
TPI1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Triosephosphate isomerase. The exact sequence is proprietary.
Protein G purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat
Western Blot
Unconjugated
PBS with No Preservative
EC 5.3.1.1, MGC88108, TIM, TPI, triosephosphate isomerase, Triose-phosphate isomerase, triosephosphate isomerase 1
Mouse
IgG2a
0.1 ml
Stem Cell Markers
Primary
7167