Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-Trpv4, Clone: 1B2.6,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABS466
Description
Specifically detects Trpv4 clone: 1B2.6 in Human, Mouse samples, and it is validated for Immunocytochemistry, Western Blotting
Transient receptor potential cation channel subfamily V member 4 is a multitransmembrane non-selective cation channel encoded by the TRPV4 (also known as CMT2C, HMSN2C, OTRPC4, SMAL, SPSMA, SSQTL1, TRP12, VR-OAC, VRL-2, VRL2, VROAC) gene and belong to the OSM9-like transient receptor potential channel (OTRPC) subfamily within the transient receptor potential (TRP)3 superfamily of channels. Ankyrin (ANK) repeats/domains are shown to be important for functional channel expression, and ANK domain mutations are found in patients suffering from Spinal muscular atrophies (SMA, also known as hereditary motor neuropathies) and hereditary motor and sensory neuropathies (HMSN). (PMID 16293632, 20037587, 20037588).
Specifications
Trpv4 | |
Monoclonal | |
Purified mouse monoclonal IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
VRL2; VROAC; TRPV4 | |
GST-tagged recombinant protein corresponding to human Trpv4. | |
100 μg | |
Signaling | |
NP_001170899 | |
Stable for one year at 2°C to 8°C from date of receipt. | |
IgG2a κ |
Immunocytochemistry, Western Blot | |
1B2.6 | |
Q9HBA0 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction