Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-TUFM, Clone: CL2243, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236752

Catalog No. NBP236752


Add to cart

Description

Description

TUFM Monoclonal antibody specifically detects TUFM in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

TUFM
CL2243
Western Blot 1:500-1:1000, Immunohistochemistry 1:5000 - 1:10000, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:5000 - 1:10000
P49411
EFTu, EF-Tu, EFTUCOXPD4, EF-TuMT, elongation factor Tu, mitochondrial, P43, Tu translation elongation factor, mitochondrial
Mouse
IgG2a
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Human
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Purified
TUFM
This antibody was developed against Recombinant Protein corresponding to amino acids:AVDTYIPVPARDLEKPFLLPVEAVYSVPGRGTVVTGTLERGILKKGDECELLGHSKNIRTVVTGIEMFHKSLERAEAGDNLGALVRGLKREDLRRGLVMVKPGSI
Protein A purified
RUO
Primary
7284
Protein A purified
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only