Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Tyrosine Hydroxylase, PE, Clone: 2D3.1G7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242212PE

Catalog No. NBP242212PE

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Specifications

Tyrosine Hydroxylase
2D3.1G7
Flow Cytometry 1 - 2.5 ug/mL, Flow (Intracellular)
P07101
DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase
Mouse
IgG1
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Rat
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
Purified
TH
Partial recombinant human Tyrosine Hydroxylase protein made to a C-terminal sequence (between amino acids 300-528) [UniProt P07101]
58 kDa
0.1 ml
Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Prostate Cancer, Tyrosine Kinases
Primary
7054
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only