Learn More
Mouse anti-TYRP1, Clone: TA99 + TYRP1/807, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP2446220.1MG
 View more versions of this product
Catalog No. NBP24462201
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry (Frozen), Immunofluorescence
TYRP1 Monoclonal antibody specifically detects TYRP1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Frozen, Immunofluorescence.
Specifications
Specifications
TYRP1
TA99 + TYRP1/807
Unconjugated
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
b-PROTEIN, CAS25,6-dihydroxyindole-2-carboxylic acid oxidase, Catalase B, CATB, EC 1.14.18, EC 1.14.18.-, EC 1.14.18.1, Glycoprotein 75, GP75DHICA oxidase, Melanoma antigen gp75, TRP-1, TRPTYRP, tyrosinase-related protein 1TRP1, TYRRP
Mouse
IgG
Protein A or G purified
RUO
Primary
7306
Human, Mouse
Western Blot, Flow Cytometry, Immunohistochemistry (Frozen), Immunofluorescence
0.2 mg/mL
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Frozen 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml
Purified
TYRP1
SK-MEL-23 cells (TA99); Recombinant full-length human TYRP1 protein (TYRP1/807)
75 kDa
0.1 mg
Store at 4C.
Monoclonal
It reacts with a 75kDa melanocyte-specific gene product, identified as Tyrosinase-related protein-1 (TRP-1). It is involved in melanin synthesis. TRP1 is present on the melanosomal membranes of melanoma, normal melanocytes and nevi. Recent evidence suggests that TRP-1 is involved in maintaining stability of tyrosinase protein and modulating its catalytic activity. TRP-1 is also involved in maintenance of melanosome ultrastructure and affects melanocyte proliferation and cell death.