Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-UACA, Clone: AE-5, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2443560.2MG
Description
Ensure accurate, reproducible results in Immunocytochemistry, Immunofluorescence
UACA Monoclonal antibody specifically detects UACA in Human samples. It is validated for Immunofluorescence, Immunocytochemistry.Specifications
UACA | |
Monoclonal | |
0.2mg/mL | |
Immunofluorescence 0.5 - 1.0 ug/ml, Immunocytochemistry 0.5 - 1.0ug/ml | |
FLJ10128, KIAA1561MGC141969, MGC141967, nuclear membrane binding protein, NUCLING, uveal autoantigen with coiled coil domains and ankyrin repeats, uveal autoantigen with coiled-coil domains and ankyrin repeats | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
This monoclonal antibody is part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the nuclear membrane expressed in human cells. It can be used to stain the nuclear membrane in cell or tissue preparations and can be used as a marker of the nuclear membrane in subcellular fractions. It produces a ring pattern around the nucleus of cells of normal and malignant cells and may be used to stain the nuclear membrane of cells in fixed or frozen tissue sections. It can be used with paraformaldehyde fixed frozen tissue or cell preparations and formalin fixed, paraffin-embedded tissue sections.,The nuclear envelope (also known as the perinuclear envelope, nuclear membrane, nucleolemma or karyotheca) is the double membrane of the nucleus that encloses genetic material in eukaryotic cells. It s | |
Store at 4C. | |
IgG1 κ |
Immunofluorescence, Immunocytochemistry | |
AE-5 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
UACA | |
Nuclei of myeloid leukemia biopsy cells | |
0.2 mg | |
Apoptosis | |
55075 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction