Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-UACA, Clone: AE-5, Novus Biologicals™

Mouse Monoclonal Antibody

$129.00 - $299.00

Specifications

Antigen UACA
Clone AE-5
Concentration 0.2mg/mL
Immunogen Nuclei of myeloid leukemia biopsy cells
Gene ID (Entrez) 55075
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP24435600 View Documents Novus BiologicalsSupplier Diversity Partner
NBP2443560.02MG
0.02 mg Each for $129.00
Add to cart
 
NBP24435601 View Documents Novus BiologicalsSupplier Diversity Partner
NBP2443560.1MG
0.1 mg Each for $299.00
Add to cart
 
NBP24435602 View Documents Novus BiologicalsSupplier Diversity Partner
NBP2443560.2MG
0.2 mg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in Immunocytochemistry, Immunofluorescence

UACA Monoclonal antibody specifically detects UACA in Human samples. It is validated for Immunofluorescence, Immunocytochemistry.
Specifications

Specifications

UACA
0.2mg/mL
55075
Monoclonal
Mouse
IgG1 Kappa
Purified
RUO
Protein A or G purified
AE-5
Nuclei of myeloid leukemia biopsy cells
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Primary
This monoclonal antibody is part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the nuclear membrane expressed in human cells. It can be used to stain the nuclear membrane in cell or tissue preparations and can be used as a marker of the nuclear membrane in subcellular fractions. It produces a ring pattern around the nucleus of cells of normal and malignant cells and may be used to stain the nuclear membrane of cells in fixed or frozen tissue sections. It can be used with paraformaldehyde fixed frozen tissue or cell preparations and formalin fixed, paraffin-embedded tissue sections.,The nuclear envelope (also known as the perinuclear envelope, nuclear membrane, nucleolemma or karyotheca) is the double membrane of the nucleus that encloses genetic material in eukaryotic cells. It separates the contents of the nucleus (DNA in particular) from the cytosol (cytoplasm). Numerous nuclear pores are present on the nuclear envelope to facilitate and regulate the exchange of materials (for example, proteins and RNA) between the nucleus and the cytoplasm. The space between the two membranes that make up the nuclear envelope is called the perinuclear space (also called the perinuclear cisterna), and is usually about 20 - 40 nm wide. Each of the two membranes is composed of a lipid bilayer. The outer membrane is continuous with the rough endoplasmic reticulum. The inner membrane is erected upon the nuclear lamina, a network of intermediate filaments made of lamin, that plays a role in mitosis and meiosis. The type of lamins present are A, B1, B2, and C. The nuclear envelope may also play a role in the disposition of chromatin inside the nucleus. The lamina acts as a site of attachment for chromosomes. It also acts like a shield for the nucleus. During prophase in mitosis, the chromatids begin condensing to form chromosomes, and the nuclear envelope begins to disintegrate. During metaphase, the nuclear envelope is completely disintegrated, and the chromosomes can be pulled apart as chromatids by the spindle fibers.
Unconjugated
Immunofluorescence, Immunocytochemistry
Store at 4C.

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit