Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-UACA, Clone: UACA/1222, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP2443570.02MG

 View more versions of this product

Catalog No. NBP24435700


Add to cart

Description

Description

Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunofluorescence

UACA Monoclonal antibody specifically detects UACA in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence.
Specifications

Specifications

UACA
UACA/1222
Unconjugated
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
FLJ10128, KIAA1561MGC141969, MGC141967, nuclear membrane binding protein, NUCLING, uveal autoantigen with coiled coil domains and ankyrin repeats, uveal autoantigen with coiled-coil domains and ankyrin repeats
Mouse
IgG1 Kappa
Protein A or G purified
RUO
Store at 4C.
Monoclonal
UACA (Uveal Autoantigen with Coiled-coil domains and Ankyrin repeats) is a 1,416 amino acid nuclear membrane protein. It was originally identified as an autoantigen in patients with panuveitis, a characteristic of Vogt-Koyanagi-Harada disease, and in patients with Graves' disease. UACA was also later identified as Nucling, an mRNA differentially expressed in F9 embryonal carcinoma cells during cardiac muscle differentiation. UACA appears to function as a pro-apoptotic protein that recruits the apaf-1-pro-caspase-9 complex for the induction of apoptosis to mediate the cell-death pathway.
Western Blot, Flow Cytometry, Immunofluorescence
0.2mg/mL
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml
Purified
UACA
Recombinant human UACA protein
160 kDa
0.02 mg
Apoptosis
Primary
55075
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only