Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2I, Mouse, Clone: 1B10, Abnova™

Mouse monoclonal antibody raised against recombinant UBE2I.

Manufacturer:  Abnova Corporation MAB10344

Catalog No. 89-108-531


Add to cart

Description

Description

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. Four alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Specifications

Specifications

UBE2I
1B10
Mouse monoclonal antibody raised against recombinant UBE2I.
In ascites (0.03% sodium azide)
Liquid
UBE2I
Recombinant protein corresponding to human UBE2I.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user.
UBE2I
C358B7.1/P18/UBC9
Mouse
IgG1
RUO
Primary
7329
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit