Learn More
Mouse anti-UCP1, Clone: 3111, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243550
Catalog No. NBP243550
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot
UCP1 Monoclonal antibody specifically detects UCP1 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
UCP1
3111
Western Blot 1:500-1:3000
Purified
UCP1
Recombinant protein encompassing a sequence within the center region of human UCP1. The exact sequence is proprietary.
33 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Mouse
Western Blot
Unconjugated
PBS with No Preservative
SLC25A7mitochondrial brown fat uncoupling protein 1, Solute carrier family 25 member 7, thermogenin, UCP, UCP 1, uncoupling protein 1 (mitochondrial, proton carrier)
Mouse
IgG1
Protein G purified
RUO
Primary
7350