Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VCAM-1/CD106 Mouse anti-Rat, Clone: MR106, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
$204.00 - $642.00
Specifications
Antigen | VCAM-1/CD106 |
---|---|
Clone | MR106 |
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry (Frozen), Western Blot, Immunoprecipitation, Flow Cytometry, Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018541
|
Novus Biologicals
NBP1433680.025MG |
0.025 mg |
Each for $204.00
|
|
|||||
NBP14336805
|
Novus Biologicals
NBP1433680.05MG |
0.05 mg |
Each for $303.00
|
|
|||||
NBP14336801
|
Novus Biologicals
NBP1433680.1MG |
0.1 mg |
Each for $461.00
|
|
|||||
NBP1433685
|
Novus Biologicals
NBP1433680.5MG |
0.5 mg |
Each for $642.00
|
|
|||||
Description
VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
VCAM-1/CD106 | |
0.5 mg/mL | |
Immunohistochemistry (Frozen), Western Blot, Immunoprecipitation, Flow Cytometry, Immunohistochemistry | |
Unconjugated | |
RUO | |
Rat | |
CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
VCAM1 | |
IgG1 κ | |
Affinity Purified |
MR106 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Mouse | |
Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
7412 | |
The immunogen for this antibody was CD106. | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'