Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV-1 Gag p24 Antibody (7F4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP241339
Description
HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot, ELISA.Specifications
HIV-1 Gag p24 | |
Monoclonal | |
1 mg/mL | |
Western Blot 0.2-0.5 μg/mL, ELISA 0.2- 0.5 μg/mL | |
Capsid protein p24, Human immunodeficiency virus 1, Human immunodeficiency virus type 1 p24 | |
Mouse | |
24, 41, 55 kDa | |
0.1 mg | |
Secondary | |
By Western blot, anti-HIV-1 Gag p24 antibody detects a ∼24 kDa, a ∼41 kDa, and a ∼55 kDa protein, corresponding to HIV-1 Gag p24 and to its precursors p41 and p55, respectively, in HIV-1 samples. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA | |
7F4 | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
gag | |
Antibody was raised against a recombinant full-length HIV-1 Gag p24 protein. Amino Acid Squence: pivqniqgqmvhqaisprtlnawvkvveekafspevipmfsalsegatpqdlntmlntvgghqaamqmlketineeaaewdrvhpvhagpiapgqmreprgsdiagttstlqeqigwmtnnppipvgeiykrwiilglnkivrmysptsildirqgpkepfrdyvdrfyktlraeqasqevknwmtetllvqnanpdcktilkalgpaatleemmtacqgvggpghkarvla | |
Protein A purified | |
RUO | |
155030 | |
Virus | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction