Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-WDR66, Clone: 2A6F7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237381

Catalog No. NBP237381


Add to cart

Description

Description

WDR66 Monoclonal antibody specifically detects WDR66 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications

Specifications

WDR66
2A6F7
Unconjugated
PBS with 0.05% Sodium Azide
FLJ39783, MGC33630, WD repeat domain 66, WD repeat-containing protein 66
Mouse
IgG1
Protein A or G purified
RUO
Primary
144406
Western Blot, Flow Cytometry, ELISA, CyTOF
1 mg/mL
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready
Affinity Purified
WDR66
Purified recombinant fragment of human WDR66 (AA: 1-250) expressed in E. Coli.
130 kDa
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only