Learn More
Mouse anti-ZFP57, Clone: 1115, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243814
Catalog No. NBP243814
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot
ZFP57 Monoclonal antibody specifically detects ZFP57 in Human samples. It is validated for Western Blot.
Specifications
Specifications
ZFP57
1115
Western Blot 1:1000-1:10000
Purified
ZFP57
Recombinant protein encompassing a sequence within the N-terminus region of human ZFP57. The exact sequence is proprietary.
52 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot
Unconjugated
PBS with No Preservative
C6orf40, TNDM1, zfp-57, zinc finger protein 57 homolog (mouse), zinc finger protein 698, ZNF698
Mouse
IgG2a
Protein A purified
RUO
Primary
346171