Learn More
Novus Biologicals™ Mouse ASK1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP269953
Description
The Mouse ASK1 ELISA Kit (Colorimetric) measures mouse ASK1 in serum, plasma and other biological fluids.Specifications
15.63 - 1000 pg/mL | |
This kit recognizes Mouse ASK1 in samples. No significant cross-reactivity or interference between Mouse ASK1 and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
4217 | |
Mouse | |
Storage of components varies. See protocol for specific instructions. |
9.38 pg/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
MAP3K5 | |
1 Kit | |
ASK1 |