Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse PD-L1/B7-H1 His-tag Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Supplier: R&D Systems™ 9048B7100
This item is not returnable.
View return policy
Description
Measured by its binding ability in a functional ELISA. When Recombinant Mouse PD-L1/B7-H1 is immobilized at 25 ng/mL (100 μL/well), the concentration of Recombinant Mouse PD-1 Fc Chimera (Catalog # 1021-PD ) that produces 50% of the optimal binding response is approximately 8-40 ng/mLSpecifications
Q9EP73 | |
Lyophilized from a 0.2 μm filtered solution in PBS. | |
M.W. (Observed): 49-62 kDa, reducing conditions; M.W. (theoretical): 26 kDa | |
Mouse myeloma cell line, NS0-derived mouse PD-L1/B7-H1 protein Phe19-His239, with a C-terminal 6-His tag | |
<0.10 EU / 1 μg of the protein by the LAL method. | |
B7-H1/PD-L1/CD274 | |
Measured by its binding ability in a functional ELISA. When Recombinant Mouse B7-H1/PD-L1 is immobilized at 25ng/mL (100μL/well), the concentration of Recombinant Mouse PD-1 Fc Chimera that produces 50% of the optimal binding response is approximately 8-40ng/mL | |
Mouse | |
Reconstitute at 400 μg/mL in PBS. | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Bioactivity | |
29126 | |
100 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
Cd274 | |
Unconjugated | |
Recombinant | |
Mouse myeloma cell line, NS0-derived Phe19-His239, with a C-terminal 6-His tag | |
PD-L1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction