Learn More
Acro Biosystems Recombinant Protein;Mouse B7-H3 / CD276 Protein, Fc Tag;HEK293;100UG;B73,B7-H3,CD276,B7 homolog 3,B7H3 DFS Item
Manufacturer:  Acro Biosystems B73M5255100UG
Recombinant Protein;Mouse B7-H3 / CD276 Protein, Fc Tag;100UG;Mouse B7-H3, Fc Tag (B73-M5255) is expressed from human 293 cells (HEK293). It contains AA Val 29 - Phe 244 (Accession # NP_598744).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.1 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-113-8284