Learn More
Novus Biologicals™ Mouse BAK ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP269931
Catalog No. NBP269931
Add to Cart
Description
Description
The Mouse BAK ELISA Kit (Colorimetric) measures mouse BAK in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
This kit recognizes Mouse BAK1 in samples. No significant cross-reactivity or interference between Mouse BAK1 and analogues was observed.
Biological Fluids, Plasma, Serum
578
Mouse
Storage of components varies. See protocol for specific instructions.
0.10 ng/mL
ELISA Kit (Colorimetric)
ELISA
BAK1
1 Kit
BAK
Product Suggestions
Product Suggestions